missing translation for 'onlineSavingsMsg'
Learn More

Titanium wire, 0.81mm (0.032in) dia x 90cm (35in) long, annealed, 99.7% (metals basis), Thermo Scientific Chemicals

Form: approx. 12.9g/pc

£112.00 - £409.00

Chemical Identifiers

CAS 7440-32-6
Molecular Formula Ti
Molecular Weight (g/mol) 47.87
MDL Number MFCD00011264,MFCD00151301
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
Synonym oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11385779
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010395.KK
10 Pc.
£112.00
Pack of 10
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11355868
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010395.KD
50 Pc.
£409.00
Pack of 50
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-32-6
47.87
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
titanium
Ti
MFCD00011264,MFCD00151301
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
CHEBI:33341
[Ti]

Specifications

Silver to Gray
Silver Gray wire
99.7% (Metals basis)
MFCD00011264,MFCD00151301
RTAQQCXQSZGOHL-UHFFFAOYSA-N
titanium
23963
Odorless
Titanium
7440-32-6
4.506g/cm3
Ti
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
[Ti]
47.87
CHEBI:33341
Wire
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-142-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only