missing translation for 'onlineSavingsMsg'
Learn More
Titanium wire, 2.0mm (0.08in) dia x 91cm (36in) long, 99.7% (metals basis), Thermo Scientific Chemicals Product Code.: 15435945

Titanium wire, 2.0mm (0.08in) dia x 91cm (36in) long, 99.7% (metals basis), Thermo Scientific Chemicals

Product Code. 15435945
5 Pc.
Click to view available options
Quantity:
25 Pc.
5 Pc.
This item is not returnable. View return policy

Product Code. 15435945

Brand: Thermo Scientific Alfa Aesar 010397.KJ

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-32-6
Molecular Formula Ti
Molecular Weight (g/mol) 47.87
MDL Number MFCD00011264,MFCD00151301
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
Synonym oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]

Specifications

Appearance Silver Gray wire
Density 4.506g/cm3
Assay Percent Range 99.7% (Metals basis)
Quantity 5 Pc.
Odor Odorless
Form Wire
Chemical Name or Material Titanium

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ18J018 Certificate TypeCertificate of Analysis Date11/01/2024 Catalog Number010397.KM,010397.KJ

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Titanium wire, 2.0mm (0.08in) dia x 91cm (36in) long, 99.7% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.