Learn More
R&D Systems™ TiterTACS In Situ Detection Kit - Colorimetric
Designed specifically for quantitation of apoptosis in suspension and monolayer cell cultures
Brand:  R&D Systems™ 4822-96-K
Additional Details : Weight : 0.29100kg
Product Code. 18451850
  • £507.55 / 96 tests
Estimated Shipment: 05-08-2024
to see stock.
Includes: Proteinase K Solution; Cytonin; TdT enzyme, dNTP mix and buffers; Strep-HRP; TACS-Sapphire; Blue Streptavidin-Diluent; Cations; TACS-Nuclease™ and buffer; Nuclear Fast Red
Description
Description
Allows quantitative colorimetric analysis of a large number of cell samples using a 96-well plate
Provides quantitation of apoptosis in cultured cells without direct counting of labeled cells
Sensitive and easy to follow assay
Specifications
Specifications
96 Tests
The product requires storage at both -20 to -70° C and 2 - 8° C. Consult the product insert for specific storage temperatures. Do not use past expiration date.