Learn More
MP Biomedicals™ Titertek™ Finnpipette™ Multistepper Pipet Tips
Hydrophobic tips with very low coefficient of friction
Brand: MP Biomedicals™ 0977943C2
132.92 GBP valid until 2024-12-31
Use promo code "21895" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifically for the Titertek™/Labsystems 8-channel Multistepper pipette
- Made from virgin polypropylene material
- Comes with robot-compatible (large-micron) hydrophobic filters to eliminate instrument contamination
- 96 tips per rack
- Volume: 1500μL
Specifications
1500 μL | |
Natural | |
Polypropylene |
Non-sterile | |
Bulk |