missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ProcartaPlex™ Automation Setup Kit

Product Code. 17489292
Click to view available options
Quantity:
6 Tests
Unit Size:
6 tests
This item is not returnable. View return policy

Product Code. 17489292

Brand: Invitrogen™ EPXSETUP384

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The ProcartaPlex Automation Setup Kit enables setup, testing, and validation of instruments to automate crucial steps of the ProcartaPlex 384-well assay protocol.

The ProcartaPlex Automation Setup Kit enables setup, testing, and validation of instruments to automate crucial steps of the ProcartaPlex 384-well assay protocol, such as liquid handling and plate washing, before running an actual assay with valuable sample. The user guide describes how to adjust and optimize an automated magnetic plate washer as needed during the ProcartaPlex 384-well assay workflow in order to ensure optimal performance and avoid bead loss.

Each kit provides six optimization tests for automated magnetic plate washing.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Basic and Setup Kits
Quantity 6 Tests
For Use With (Equipment) FLEXMAP 3 D™ INTELLIFLEX™
Product Line ProcartaPlex
Content And Storage

• Automation Setup Beads (1X)
• 1 x 200 mL Wash Buffer (10X)
• 1 Bottle Reading Buffer (1X)
• Adhesive film
• ProcartaPlex 384-well flat bottom plate

Store at 2°C to 8°C.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.