Learn More
Heidolph Instruments Titramax 1000, EU Plug
Titramax 1000 is the largest model of the series and contains all the features of the Titramax 100.
Brand: Heidolph Instruments 544-12200-06
1213.25 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Additional Details : Weight : 8.26000kg
Description
- The largest model of the Titramax series contains all the features of the Titramax 100: a shaking orbit of 1.5 mm, speeds of up to 1350 rpm, and a timer.
- With a storage capacity of up to six multi-well plates, a higher number of samples can be processed. The option to integrate the Titramax 1000 into the modular incubation system also allows temperature-controlled processes to be carried out without problems.
- With this model, easy loading and removal of the plates can be carried out comfortably due to the recess and the spacings in the rubber mat.
Specifications
1.5 mm | |
230 V | |
Titramax 1000 |
0 to 1350 rpm | |
UK Plug |