Learn More
SI Analytics™ Titration Vessel
SI Analytics™ Titration Vessel is a temperature controlled vessel that features a double jacket with 5 openings.
Brand:  SI Analytics™ 285213499
Product Code. 10672832
  • £1295.00 / Each
Estimated Shipment: 14-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Temperature-controlled titration vessel
  • Double jacket
  • 4 openings NS 14.5 and 1 opening NS 7.5
  • Includes a micro-titration tip with PTFE insert and gas supply tube
  • Micro design
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Titration Vessel
Micro titration tip with PTFE insert and gas supply tube
4 x NS 14.5, 1 x NS 7.5
3 to 10 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
SI Analytics™ Titration Vessel > Capacity: 3 To 10mL