missing translation for 'onlineSavingsMsg'
Learn More

SI Analytics™ TitroLine™ 5000 Automatic Titrator

Easily obtain precise and quick results without the need for training or knowledge of automatic titration. SI Analytics™ TitroLine™ 5000 Automatic Titrator is the first choice if you are looking for an easy to use automatic titrator for any application.

Brand:  SI Analytics™ 285225770

 View more versions of this product

Product Code. 15362367

  • £3015.00 / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket

Includes:
Cat. No. 15314408 - TitroLine 5000 with 20mL dosing unit
Ready to use assembled 20mL dosing unit, manual controller, titration clamp, stand rod, magnetic stirrer TM 50 and power supply 100 to 240V

Cat. No. 15324408 - TitroLine 5000 with 50mL dosing unit
pH electrode and buffer set, with ready to use assembled 50mL dosing unit, manual controller, titration clamp, stand rod, magnetic stirrer TM 50 and power supply 100 to 240V
This item is not returnable. View return policy

Description

Description

  • High resolution pH/mV-measurement input for pH-, ORP-, silver - and further mV-electrodes
  • pH/mV-input with electrode socket according to DIN 19 262 or additional with BNC socket insert (Z 860)
  • Pt 1000 and NTC 30 temperature measurement input (socket 2 x 4mm) for automatic temperature compensation
  • Pre-installed standard methods for FOS/TAC, alkalinity, total acidity in drinks, chloride
  • Unit operated using the keys on the device itself, the controller TZ 3880 and optional PC-keyboard (USB)
  • Graphics-capable TFT display
  • Linear and dynamic titration to equivalence points
  • Titration to pH and mV-ends points
  • Stirrer Connection: TM 50 power supply directly through piston burette
  • High resolution pH/mV-measurement input for pH-, ORP-, silver - and further mV-electrodes
  • Same manual titration and dosing function as the piston burette Titronic™ 300
  • Typical applications:
    • pH-value, alkalinity (p+m-value)
    • FOS/TAC
    • Total Kjeldahl nitrogen
    • Permanganate index and COD
    • Chloride in wastewater

Certifications

CE-mark: EMC: 2004/108/EG; safety EG- Directive 2006/95

Compliance

ISO 8655, part 6

TRUSTED_SUSTAINABILITY
Specifications

Specifications

ISO 8655, part 6, CE-mark: EMC: 2004/108/EG; safety EG- Directive 2006/95
Automatic Titrator
Automatic Titrators
Basic unit without electrode, with ready to use assembled 50 ml dosing unit, manual controller, titration clamp, stand rod, magnetic stirrer TM 50 and power supply 100-240 V
Display resolution: 0.005 to 0.025mL; Burette resolution: 8000
13.5 cm
100 to 240V; 50/60Hz, power input 30 VA
50/60 Hz
1 x USB-A and 1 x USB-B, 2 x RS-232-C
Graphics-capable TFT
Determine chloride in wastewater, permanganate index and COD, total Kjeldhal nitrogen, FOS/TAC, pH value, alkalinity
TL 5000/50 M1
10°C to 40°C
20.5 cm
100 to 240v
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.