Learn More
SI Analytics TitroLine™ TL 7000-M2/20
Features the highest safety for measuring and calibrating through wireless sensor recognition for ID-electrodes. SI Analytics™ TitroLine™ 7000 Basic Unit Model TL 7000-M2/20 possesses general titration feautres but offers improved performace.
Brand:  SI Analytics 285220170
Product Code. 13446979
  • £5915.00 / Each
Explore more special offers
Description
Description
  • The brilliant TFT display allows perfect readout even from the side
  • The intelligent exchangeable units are storing all relevant reagent and unit data
  • The automatic wireless recognition of SCHOTT™ Instruments
  • ID electrodes guarantees accurate calibration and measurements
  • 3 USB and 2 RS-232 ports allow to connenct e.g. USB keyboards, USB printers, USB storage media, stirrer, balance, PC and other
  • SI Analytics instruments
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TitroLine 7000 Basic Unit
Polypropylene
10°C to 40°C
15.3 cm
Brown Glass Bottle For Titrant, GL 45 And S 40-Bottle Adapter, Hoses, Drip Off Tube And Titration Tip. With pH-Combination Electrode IL-pHT-A120MF-DIN-N And Buffer Set
TL 7000-M2/20
45 cm
29.6 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Guides
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
SI Analytics TitroLine™ TL 7000-M2/20 > WidthMetric: 15.3cm