missing translation for 'onlineSavingsMsg'
Learn More

SI Analytics™ TitroLine™ 7500 Volumetric Titrators

Easy and precise, with standard methods for different applications. SI Analytics™ TitroLine™ 7500 Volumetric Titrator offers more performance and is the volumetric generalist for a wide range of use.

Brand:  SI Analytics™ 285220820

5201.25 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 13486768

  • £6000.00 / Each
Estimated Shipment: 14-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Fast, easy and precise
  • With standard methods for different applications (titer determination, blank value…)
  • High visible full color display, that can be easily viewed from a distance and extreme angles
  • Storage of results via USB port (PDF and CSV -format)
  • With intelligent interchangeable modules (only TitroLine™ 7500 KF)
  • Applications for the TitroLine 7500™ KF: KF volumetry, dead-stop-titrations (SO2, bromine number)
 
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DIN EN ISO 8655
KF volumetry, dead-stop-titrations (SO2,bromine number).
Polypropylene
10°C to 40°C
15.3 cm
100 to 240v
Volumetric KF-Titrator
Interchangeable Module WA 10
TL 7500 KF 10
45 cm
29.6 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.