missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SI Analytics™ TitroLine™ 7750
SI Analytics™ TitroLine™ 7750 is the all-rounder for both potentiometric titration and volumetric KF titration.
£4990.00 - £7695.00
Specifications
Material | Polypropylene |
---|---|
Model | TitroLine™ 7750 |
Range (pH) | -3 to 18 |
Temperature Range (Metric) Ambient | 10°C to 40°C |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
13496768
|
SI Analytics™
285220250 |
Stand, TZ 1510, electrode clamp Z 305, hand controller TZ 3880 |
£4990.00
Each |
Estimated Shipment: 09-10-2024
Log in to see stock. |
|||||
15524923
|
SI Analytics™
285220950 |
KF accessories: basic titrator unit, exchange unit WA 20, TM 235 KF titration stand with integrated stirrer and pump, titration vessel TZ 1770, micro double platinum electrode KF 1100 and starter kit |
£7695.00
Each |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||||
Description
- pH/mV-input with 24 bit transducer Electrode socket according to DIN 19 262 or additional with BNC socket insert RFID receiver for SI Analytics ID electrodes
- Connector (μA) for double platinum electrodes Polarisation voltage adjustable from 40 to 220 mV
- Highly visible full color display, that can be easily viewed from a distance and extreme angles With new interchangeable modules which all relevant reagent and unit data can be stored
- Expandable thanks to the 2 x USB-host, 1 x USB-PC, 1 x LAN and 2 x RS232 ports
- Connectable are e.g. USB keyboard, USB printer, barcode reader, USB flash drives, balances, PC and further devices such as piston burettes and sample changers
- Storage of results via USB or LAN connection, including method transfer
Specifications
Polypropylene | |
TitroLine™ 7750 | |
-3 to 18 | |
10°C to 40°C |