Learn More
SI Analytics™ TitroLine™ 7800 Series Titrators
The TitroLine™ 7800 is a universal titrator with digital sensor technology featuring enhanced methods for potentiometric and KF titration
Brand:  SI Analytics™ 285220990
5153.75 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Product Code. 15564923
  • £5955.00 / Each
Estimated Shipment: 13-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Universal potentiometric titrator with analogue and IDS digital sensor technology for the determination of pH and ORP values
  • Exceptional high visibility colour graphic display for viewing even at extreme angles
  • Clear graphic representation of titration curves
  • Pre-installed standard methods for potentiometric and KF titration
  • USB connectivity
  • Ideally suited for water analysis
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50 user methods
3
USB export or pdf/CSV format
Online display
English
TitroLine 7800
0 to 14
0.004 pH units
153mm x 296mm x 450mm
11.6 in.
1
1xLAN, 2xUSB-A, 1xUSB-B, 2xRS232
Color VGA TFT display with 320x240 pixels
One analogue and one digital (IDS) measuring input
Polypropylene Housing
1200 mV +\-
0.2mV
5/10/20/50 mL
6.0 in.