Learn More
SI Analytics Titronic 300 Module 2 TM 300/50 with stirrer
Motor-driven piston burette with 20 or 50 ml dosing unit ideal for manual titrations and highly precise dosing tasks.
Brand:  SI Analytics 285225830
Product Code. 15341645
  • £2090.00 / Each
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Motor-driven piston burette with 20 or 50 ml dosing unit
  • For manual titrations and highly precise dosing tasks
  • The TITRONIC ™ 300 can completely controlled from a PC via the USB- or RS-232-interface
  • Up to 16 burettes can be connected to one another via the RS232 interface according to the daisy chain principle
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Titronic 300 Module 2 TM 300/50 100-240V with stirrer
Graphics-capable TFT display.
Stirrers
100-240V; 50/60 Hz