Learn More
TL1A Polyclonal Antibody, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
£171.00 - £1422.00
Specifications
Antigen | TL1A |
---|---|
Concentration | 0.1-1.0 mg/mL |
Applications | ELISA, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17845673
|
Invitrogen
500-P240-50UG |
50μg |
£171.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17835523
|
Invitrogen
500-P240-100UG |
100 μg |
£220.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17855783
|
Invitrogen
500-P240-1MG |
1 mg |
£1422.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AA Sequence of recombinant protein: QLRAQGEASV QFQALKGQEF APSHQQVYAP LRADGDKPRA HLTVVRQTPT QHFKNQFPAL HWEHELGLAF TKNRMNYTNK FLLIPESGDY FIYSQVTFRG MTSECSEIRQ AGRPNKPDSI TVVITKVTDS YPEPTQLLMG TKSVCEVGSN WFQPIYLGAM FSLQEGDKLM VNVSDISLVD YTKEDKTFFG AFLL. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human TL-1A. Anti-Human TL-1A-specific antibody was purified by affinity chromatography employing an immobilized Human TL-1A matrix. Sandwich ELISA: To detect Human TL-1A by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human TL-1A (500-P240Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human TL-1A. Western Blot: To detect Human TL-1A by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human TL-1A is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
The protein encoded by this gene is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. This protein is abundantly expressed in endothelial cells, but is not expressed in either B or T cells. The expression of this protein is inducible by TNF and IL-1 alpha. This cytokine is a ligand for receptor TNFRSF25 and decoy receptor TNFRSF21/DR6. It can activate NF-kappaB and MAP kinases, and acts as an autocrine factor to induce apoptosis in endothelial cells. This cytokine is also found to inhibit endothelial cell proliferation, and thus may function as an angiogenesis inhibitor. An additional isoform encoded by an alternatively spliced transcript variant has been reported but the sequence of this transcript has not been determined.Specifications
TL1A | |
ELISA, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
O95150 | |
9966 | |
E.coli-derived Recombinant Human TL-1A | |
Antigen affinity chromatography | |
TNFSF15 |
0.1-1.0 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
PBS with no preservative | |
bM20K13.3 (tumor necrosis factor (ligand) superfamily, member 15); MGC129934; MGC129935; TL1; TL1A; TNF ligand-related molecule 1; TNF superfamily ligand TL1A; TNFSF15; TNLG1B; tumor necrosis factor (ligand) superfamily member 15; tumor necrosis factor (ligand) superfamily, member 15; tumor necrosis factor ligand 1B; tumor necrosis factor ligand superfamily member 15; Tumor necrosis factor ligand superfamily member 15, membrane form; Tumor necrosis factor ligand superfamily member 15, secreted form; tumor necrosis factor superfamily member 15; vascular endothelial cell growth inhibitor; vascular endothelial growth inhibitor; vascular endothelial growth inhibitor-192A; Vegi; VEGI192A | |
TNFSF15 | |
Primary | |
-20°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.