Learn More
Macherey-Nagel™ TLC Micro Kit for Beginners
TLC introductory kit for beginners, science education and method development
Brand:  Macherey-Nagel™ 814000
Product Code. 12460087
  • £281.00 / Each
Explore more special offers
Description
Description
TLC Micro-Set A for beginners.
This Micro-Set A contains all chemicals and accessories for the following experiments:
  • Separation of a mixture of fat-soluble / lipophilic dyes (test dye mixture 1) butter yellow, indophenol, sudan blue II, sudan red G
  • Separation of a mixture of anthraquinone dyes (test dye mixture 2) blue 1, blue 3, green, green blue, red, violet 1, violet 2
  • Separation of a mixture of food dyes (test dye mixture 3) brilliant black BN (E151), fast red E, erythrosine (E127), yellow orange S (sunset yellow CFC, E110), naphthol red S, ponceau 4 R (E124), tartrazine (E102)
  • Separation of dyes from felt tip pens.
This product contain harmful substances which must be specially labeled as hazardous. For detailed information please see SDS.
Specifications
Specifications
Science Education
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • TLC Micro Set A
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Flammable liquid Category 3
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contains : ammonia solution, 2-propanol, ethanol, toluene, cyclohexane