missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR4 Mouse anti-Human, Mouse, Clone: 3G9A4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66350-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
TLR4 is a member of the Toll like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. This receptor is most abundantly expressed in placenta, and in myelomonocytic subpopulation of the leukocytes. It has been implicated in signal transduction events induced by lipopolysaccharide (LPS) found in most gram-negative bacteria.Specifications
TLR4 | |
Monoclonal | |
0.77 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O00206, Q9QUK6 | |
TLR4 | |
TLR4 Fusion Protein Ag13857 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot | |
3G9A4 | |
Unconjugated | |
TLR4 | |
ARMD10, CD284, hToll, TLR4, TOLL, toll like receptor 4 | |
Mouse | |
Protein A | |
RUO | |
21898, 7099 | |
-20°C | |
Liquid |