missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLX3 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£119.00 - £295.00
Specifications
Antigen | TLX3 |
---|---|
Concentration | 300 μg/mL |
Applications | Immunoprecipitation, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16877344
|
Proteintech
17454-1-AP-20UL |
20 μL |
£119.00
20µL |
Estimated Shipment: 01-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16867344
|
Proteintech
17454-1-AP-150UL |
150 μL |
£295.00
150µL |
Estimated Shipment: 01-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The TLX3 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human TLX3. This antibody recognizes human, mouse, rat antigen. The TLX3 antibody has been validated for the following applications: IP, ELISA analysis.
Specifications
TLX3 | |
Immunoprecipitation, ELISA | |
Unconjugated | |
Human, Rat, Mouse | |
IgG |
300 μg/mL | |
Polyclonal | |
Rabbit | |
30012 | |
Primary |