Learn More
Cytiva TMB Substrate
Use for in-house immunoassays that use an HRP-conjugated antibody. Cytiva Life Sciences™ TMB Substrate use the same formulation used in the majority of Biotrak™ colorimetric immunoassays and is supplied ready to use.
Brand: Cytiva RPN4910
Code : GE
Additional Details : Weight : 0.22600kg
Description
- Suitable for in-house immunoassays that use an HRP-conjugated antibody
- Supplied ready to use
Specifications
5 Bottles | |
Liquid | |
TMB Substrate |
2°C to 8°C | |
For colorimetric Biotrak assays |