missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TMCS Silylation Reagent

Catalyze the derivatization of difficult-to-silylate compounds using Thermo Scientific™ TMCS Silylation Reagent.

Brand:  Thermo Scientific™ TS-88530

317.21 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 11562391

  • £319.59 / 25g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Catalyze the derivatization of difficult-to-silylate compounds using Thermo Scientific™ TMCS Silylation Reagent. It is an excellent adjunct for forming TMS ethers for GC determinations. TMCS is also useful for preparing TMS derivatives of organic acids.

  • Excellent adjunct for forming trimethylsilyl ethers for GC determinations
  • Used to prepare TMS derivatives of organic acids
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GC
98 to 100%
25 g
Flammable, Toxic
Room Temperature
Chlorotrimethylsilane
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ TMCS Silylation Reagent > 25g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.