missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM110 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA544449
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: MQGPAGNASR GLPGGPPSTV ASGAGRCESG ALMHSFGIFL QGLLGVVAFS Sequence homology: Human: 100%; Rabbit: 100%; Rat: 86%
TMEM110 Polyclonal antibody specifically detects TMEM110 in Human samples. It is validated for Western Blot.Specifications
TMEM110 | |
Polyclonal | |
Unconjugated | |
STIMATE | |
1810038N08Rik; 2310014H19Rik; AW554125; STIM-activating enhancer; STIM-activating enhancer encoded by TMEM110; STIMATE; Store-operated calcium entry regulator STIMATE; Tmem110; Transmembrane protein 110 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
375346 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q86TL2 | |
STIMATE | |
Synthetic peptide directed towards the N-terminal of human TMEM110 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |