missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM115, Mouse anti-Human, Clone: 3D4, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00011070-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MQRALPGARQHLGAILASASVVVKALCAAVLFLYLLSFAVDTGCLAVTPGYLFPPNFWIWTLATHGLMEQHVWDVAISLTTVVVAGRLLEPLWGALELLIFFSVVNVSVGLLGAFAYLLTYMASFNLVYLFTVRIHGALGFLGGVLVALKQTMGDCVVLRVPQVRVSVMPMLLLALLLLLRLATLLQSPALASYGFGLLSSWVYLRFYQRHSRGRGDMADHFAFATFFPEILQPVVGLLANLVHSLLVKVKICQKTVKRYDVGAPSSITISLPGTDPQDAERRRQLALKALNERLKRVEDQSIWPSMDDDEEESGAKVDSPLPSDKAPTPPGKGAAPESSLITFEAAPPTLSpecifications
TMEM115 | |
Monoclonal | |
Unconjugated | |
transmembrane protein 115 | |
PL6 | |
Mouse | |
100 μg | |
Primary | |
Human | |
Antibody |
ELISA, Western Blot | |
3D4 | |
In 1x PBS, pH 7.4 | |
BC017367 | |
TMEM115 | |
PL6 (AAH17367, 1 a.a. ∼ 351 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
11070 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction