Learn More
TMEM192 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28263-1-AP-150UL
This item is not returnable.
View return policy
Description
The TMEM192 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TMEM192. This antibody recognizes human, mouse antigen. The TMEM192 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
TMEM192 | |
Polyclonal | |
Unconjugated | |
150 μL | |
201931 | |
IgG |
Western Blot, Immunohistochemistry, ELISA | |
367 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |