missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM30C Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538863
This item is not returnable.
View return policy
Description
Specifications
TMEM30C | |
Polyclonal | |
Unconjugated | |
TMEM30CP | |
CDC50C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
644444 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
A0ZSE6 | |
TMEM30CP | |
A synthetic peptide derived from the internal region of human TMEM30C | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |