missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMOD3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12521-1-AP-20UL
This item is not returnable.
View return policy
Description
Blocks the elongation and depolymerization of the actin filaments at the pointed end. The Tmod/TM complex contributes to the formation of the short actin protofilament, which in turn defines the geometry of the membrane skeleton.Specifications
TMOD3 | |
Polyclonal | |
Unconjugated | |
TMOD3 | |
TMOD3, Tropomodulin 3, tropomodulin 3 (ubiquitous), U Tmod, Ubiquitous tropomodulin, UTMOD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29766, 300838, 50875 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9JHJ0, Q9NYL9 | |
TMOD3 | |
TMOD3 Fusion Protein Ag3215 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |