Learn More
Macherey-Nagel™ TMSH
Brand: Macherey-Nagel™ 701520.101
156.68 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Specifications
TMSH | |
1 mL |
Safety and Handling
- TMSH, 0,2 M, 10x1 mL
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : methanol, TMSH
Your input is important to us. Please complete this form to provide feedback related to the content on this product.