missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TMSI Silylation Reagent

Employ the strongest silylator available for carbohydrates and steroids with the Thermo Scientific™ TMSI Silylation Reagent.

Brand:  Thermo Scientific™ TS-88623


 View more versions of this product

Product Code. 11737027

  • £208.00 / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Employ the strongest silylator available for carbohydrates and steroids with the Thermo Scientific™ TMSI Silylation Reagent. This silylation reagent reacts quickly with hydroxyls and carboxylic acids but not with amines, and is particularly well suited in multiderivatization schemes for compounds containing both hydroxyl and amine groups. TMSI may be used to derivatize alcohols, phenols, organic acids, hormones, glycols, nucleotides, and narcotics and is excellent for C1–C5 fatty acids in serum and urine.

  • Reacts quickly and smoothly with hydroxyls and carboxylic acids but not with amines
  • Especially useful in multiderivatization schemes for compounds containing both hydroxyl and amine groups
  • Used in the derivatization of alcohols, phenols, organic acids, steroids, hormones, glycols, nucleotides and narcotics
  • Excellent for C1 through C5 fatty acids in serum and urine
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GC/MS
98 to 100%
2°C to 8°C
1-(Trimethylsilyl)-1 H-imidazole
Flammable, Irritant
10 x 1 mL ampules
10 x 1 mL ampules
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • TMSI, 10 x 1 mL Ampules N-Trimethylsilylimidazole

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P370+P378-In case of fire: Use to extinguish.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.