missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TMT11-131C Label Reagent

Product Code. 15722695 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 x 5 mg
1 x 5 mg (per tag)
3 x 0.8 mg
3 x 0.8 mg (per tag)
Unit Size:
2.4mg
5mg
Pack of 3
This item is not returnable. View return policy

Product Code. 15722695

Brand: Thermo Scientific™ A34808

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tandem mass tag (TMT) reagents are specially designed to enable identification and quantitation of proteins in different samples using tandem mass spectrometry.

Tandem mass tag (TMT) reagents are specially designed to enable identification and quantitation of proteins in different samples using tandem mass spectrometry. The TMT11-131C tag structure is identical to the other TMT10pex tags, but the TMT11-131C mass reporter is labeled with only 13C isotopes. The addition of the TMT11-131C tag enables separation of the 131-mass reporter ion of the original TMT10plex reagents into N and C variants, creating an 11th channel for relative quantitation using high resolution Orbitrap MS instruments.

The advantages of TMT label reagents include increased multiplexed relative quantitation, increased sample throughput, and fewer missing quantitative values among samples. TMT10plex label reagents plus TMT11-131C is ideal for analysis of multiple protein samples from inhibitor dose response experiments, time course experiments, or to improve statistics on biological replicates.

Features of this TMT11-131C Label Reagent include:
Multiplex—concurrent MS analysis of up to 11 samples derived from cells, tissues, or biological fluids
Robust—increased multiplex capability results in fewer missing quantitative values among samples and higher confidence among replicates
Efficient—amine-reactive, NHS-ester-activated reagents ensure efficient labeling of all peptides regardless of protein sequence or proteolytic enzyme specificity
Compatible—optimized for use with high resolution Thermo Scientific Orbirtrap MS/MS platforms, with data analysis fully supported by Proteome Discoverer 2.3

TMT11-131C is available as a standalone tag (Cat. No. A34807, A37724) or as part of this TMT10plex Isobaric Label Reagent Set plus TMT11-131C. When combined with the industry-leading, high resolution Orbitrap instruments and software, TMT reagents provide integrated total solutions for quantitative protein expression analysis.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Isobaric Label Reagent
Product Line TMT10plex
Content And Storage 11 vials, store at –5 to –30°C
Label or Dye Tandem Mass Tag (TMT) Label
Shipping Condition Dry Ice
Starting Material Peptides
Detection Method Mass Spectrometry
Workflow Step Peptide Labeling
Quantity 1 x 5 mg (per tag)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.