missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TMTpro™ 16plex Label Reagent Set

Product Code. 16426210 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 x 0.5 mg (per tag)
1 x 5 mg (per tag)
6 x 0.5 mg (per tag)
Unit Size:
5mg
Each
This item is not returnable. View return policy

Product Code. 16426210

Brand: Thermo Scientific™ A44521

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

The Thermo Scientific TMTpro 16plex Label Reagent Set enables multiplexing of up to 16 samples for multiplexed protein identification and quantitation using tandem mass spectrometry (MS/MS).

TMTpro label reagents are the next generation of tandem mass tags, designed to increase multiplexing without compromising protein identification and quantitation. The structures of the TMTpro and TMT tags are similar in being isobaric and amine reactive, but the TMTpro tag has a longer spacer region and isobutyl proline mass reporter. After fragmentation, each TMTpro tag generates a unique reporter mass (i.e., TMTpro 126-134Da) in the low-mass region of the high-resolution MS/MS spectrum used for multiplexed relative quantitation of protein expression.

The advantages of TMTpro tags include increased multiplexed relative quantitation, increased sample throughput, and fewer missing quantitative values among samples. With these capabilities, the TMTpro 16plex label reagents are suited for applications requiring higher multiplexing and fewer missing values, such as inhibitor dose response experiments, time course experiments, thermal shift assays, or single cell analysis. When combined with the industry leading, high-resolution Orbitrap instruments and software, TMTpro reagents provide integrated total solutions for quantitative protein expression analysis.

Features of TMTpro 16plex Label Reagent Sets include:
Multiplex—concurrent MS analysis of up to 16 samples derived from cells, tissues, or biological fluids
Robust—increased multiplex capability results in fewer missing quantitative values among samples and higher confidence among replicates
Efficient—amine-reactive, NHS ester-activated reagents ensure efficient labeling of all peptides regardless of protein sequence or proteolytic enzyme specificity
Compatible—optimized for use with high resolution Thermo Scientific Orbitrap MS/MS platforms, such as the Q Exactive and Orbitrap Fusion Tribrid instrument series, including the Orbitrap Eclipse Tribrid and Ascend and Orbitrap Exploris 480 mass spectrometers with data analysis fully supported by Proteome Discoverer 2.3

For high resolution analysis of TMTpro-labeled peptides, the recommended LC column for the Nanospray Flex source is the Acclaim PepMap 100 C18 LC Column (Cat. No. 164942 or 164939). For the EASY-Spray source, the recommended LC column is the EASY-Spray C18 LC Column (Cat. No. ES902 or ES003).

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Mass Spectrometer
Product Type Isobaric Label Reagent Set
Product Line TMT
Content And Storage • TMTpro-126 Label Reagent, 1 x 0.5 mg
• TMTpro-127 N Label Reagent, 1 x 0.5 mg
• TMTpro-127 C Label Reagent, 1 x 0.5 mg
• TMTpro-128 N Label Reagent, 1 x 0.5 mg
• TMTpro-128 C Label Reagent, 1 x 0.5 mg
• TMTpro-129 N Label Reagent, 1 x 0.5 mg
• TMTpro-129 C Label Reagent, 1 x 0.5 mg
• TMTpro-130 N Label Reagent, 1 x 0.5 mg
• TMTpro-130 C Label Reagent, 1 x 0.5 mg
• TMTpro-131 N Label Reagent, 1 x 0.5 mg
• TMTpro-131 C Label Reagent, 1 x 0.5 mg
• TMTpro-132 N Label Reagent, 1 x 0.5 mg
• TMTpro-132 C Label Reagent, 1 x 0.5 mg
• TMTpro-133 N Label Reagent, 1 x 0.5 mg
• TMTpro-133 C Label Reagent, 1 x 0.5 mg
• TMTpro-134 N Label Reagent, 1 x 0.5 mg

Store below -20°C.

Label or Dye Tandem Mass Tag™ (TMT) Labels
Shipping Condition Dry Ice
Starting Material Peptides
Final Product Type Peptide
Detection Method Mass Spectrometry
Workflow Step Protein Labeling
Quantity 1 x 0.5 mg (per tag)
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.