Learn More
Invitrogen™ TNF alpha Human ProQuantum Immunoassay Kit
Invitrogen™ ProQuantum™ Immunoassay Kit measures proteins with no wash steps, small sample consumption, high sensitivity, and large dynamic range on a qPCR instrument
Brand: Invitrogen™ A356015
2533.17 GBP valid until 2024-12-31
Use promo code "21989" to get your promotional price.
Additional Details : Weight : 0.50000kg
Description
- High-sensitivity: Detect low levels of proteins with greater sensitivity than traditional methods like ELISA
- Small sample consumption: Use 2 to 5 μL of sample (e.g., 2 μL vs 75 μL for triplicate wells with other methods)
- Fast, easy workflow: No wash steps, 2 hours from sample to answer
- Broad dynamic range: 5 logs minimizes sample dilutions to ensure falling within the range
- No proprietary instrument to purchase: Runs on any real-time PCR instrument
- ProQuantum software available: Robust data analysis and statistical group wise comparison
Specifications
0.0128 to 5000 pg/mL | |
Taqman™ Real-time PCR | |
Human | |
qPCR Instruments | |
Antibody-conjugate A, Antibody-conjugate B, Antibody-conjugate dilution buffer, Assay dilution buffer, Ligase, Master Mix, Standard | |
5 x 96 Reactions | |
2 to 5 μL | |
TNF alpha |
< 0.01 pg/mL | |
Proximity-based Immunoassay | |
Protein quantitation | |
1 hr. | |
FAM™ | |
RUO | |
Frozen at -20°C | |
2 hr. |