Learn More
Tobramycin, MedChemExpress
MedChemExpress Tobramycin (Nebramycin Factor 6) is a parenterally administered, broad spectrum aminoglycoside antibiotic that is widely used in the treatment of moderate to severe bacterial infections due to sensitive organisms.

£72.60 - £143.99
Specifications
Chemical Name or Material | Tobramycin |
---|---|
CAS | 32986-56-4 |
Color | Off-White |
Purity Grade Notes | Research |
Molecular Formula | C18H37N5O9 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16478306
![]() |
Medchem Express
HY-B0441 |
100 mg |
£72.60
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16462136
![]() |
Medchem Express
HY-B0441 |
500 mg |
£143.99
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tobramycin (Nebramycin Factor 6) is a parenterally administered, broad spectrum aminoglycoside antibiotic that is widely used in the treatment of moderate to severe bacterial infections due to sensitive organisms.Specifications
Tobramycin | |
Off-White | |
C18H37N5O9 | |
H2O : ≥ 100 mg/mL (213.90 mM) ∣DMSO : 2 mg/mL (4.28 mM; Need ultrasonic) | |
467.51 | |
98.0% | |
Solid |
32986-56-4 | |
Research | |
Nebramycin Factor 6 Deoxykanamycin B | |
O[C@@H]([C@@H]1O[C@@]([C@@H](C[C@@H]2O)N)([H])O[C@@H]2CN)[C@H]([C@@H](C[C@@H]1N)N)O[C@@]([C@@H]([C@@H](N)[C@@H]3O)O)([H])O[C@@H]3CO | |
467.51 | |
Research | |
COVID-19-immunoregulation |
Safety and Handling
H361
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.