Learn More
Thermo Scientific™ TOC Certified Containers
Trust our low-level certified vials for Total Organic Carbon testing and sampling. TOC-certified products are manufactured, packaged and tested to exacting standards.
Brand:  Thermo Scientific™ 40C-TOC
 View more versions of this product
Product Code. 15163488
  • £446.00 / Pack of 72
Estimated Shipment: 17-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Trust our low-level certified vials for Total Organic Carbon testing and sampling. Manufactured, tested and packaged by exacting standards,Thermo Scientific™ TOC Certified Containers are certified to contribute <10ppb or < 20ppb TOC as background.
Containers:
  • Available in several sizes and styles of containers, including the popular 40mL autosampler vials cleaned, certified and ready to use
  • Include Certificate of Analysis
Includes:
  • Sampling and storage containers
  • Autosampler compatible vials
  • Culture tubes
  • Low TOC water
Recommended for:
  • TOC testing
  • USP method 643 testing
  • Off-line grab sampling of high purity water
  • Storage of validation standards.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear glass vial with closure, dust covers, Certified <20ppb TOC w/COA
1.125 in.
28.58 mm
40 mL
72
40 mL
72 Cs.
Container
PTFE/Silicone Septa
Clear
Glass
3.75 in.
1.35 oz.
95.25 mm
Tested and certified to contribute < 20 ppb; includes Certificate of Analysis (COA)
24-414