Learn More
Tocris Bioscience™ 2-Furoyl-LIGRLO-amide
Potent and selective PAR2 agonist
Brand:  Tocris Bioscience™ 3015/1
114.21 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18075272
  • £146.00 / 1mg
Explore more special offers
Description
Description
2-Furoyl-LIGRLO-amide is a potent and selective PAR2 receptor agonist (pD2 = 7.0). Causes a dose-dependent relaxation of murine femoral arteries.
Specifications
Specifications
0.95