missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Hexa-D-arginine
Furin inhibitor
Brand: Tocris Bioscience™ 4711/1
Additional Details : Weight : 0.00970kg
Description
Hexa-D-arginine is an inhibitor of furin (Ki values are 0.106, 0.58 and 13.2 μM for furin, PACE4 and PC1 respectively).Specifications
5045 | |
Store at -20°C | |
M.W. (Observed): 954.16 |
95% | |
Dibasic-processing enzyme, EC 3.4.21, EC 3.4.21.75, FUR, FURdibasic processing enzyme, furin (paired basic amino acid cleaving enzyme), furin, membrane associated receptor protein, PACE, PACEFES upstream region, paired basic amino acid cleaving enzyme (furin, membrane associated receptorprotein), Paired basic amino acid residue-cleaving enzyme, PC, PCSK3, PCSK3furin, proprotein convertase subtilisin/kexin type 3, SPC1 | |
1 mg |