Learn More
Tocris Bioscience™ Metastin (human)
Potent, endogenous ligand for kisspeptin receptor
Brand:  Tocris Bioscience™ 1443/100U
191.23 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18015362
Description
Description
Metastin (human) is a potent endogenous ligand of the kisspeptin receptor (KISS1, GPR54). Binds with high affinity to rat and human KISS1 receptors with Ki values of 1.80 and 1.45 nM respectively. Inhibits chemotaxis, invasion and metastasis of human melanomas and breast carcinomas. Stimulates gonadotropin secretion following i.c.v. administration.
Specifications
Specifications
95%
Store at -20°C