missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Riedel-de Haën Toluene
Click to view available options
Quantity:
7 L
Description
- ACS general use solvents.
- Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis.
- Suitable for less demanding applications such as routine isocratic HPLC.
- Variations and decrease issues traceable to solvent contamination.
- All solvents are tested to meet ACS general use specifications.
Specifications
pH | No data available |
Melting Point | -95°C |
Color | Colorless |
CAS Max % | 100.00% |
Density | 0.867 g/cm3 (20°C) |
Boiling Point | 110.6°C |
Quantity | 7 L |
Flash Point | 7.1 %(V) |
Packaging | Returnable Stainless Steel Can |
Physical Form | Liquid |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
Product Identifier
- Toluene
- Danger
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- Reproductive toxicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'