missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toluene, 99.5%, ACS reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mL
500 mL
1 L
2.5 L
25 L
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
Metal Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -95.0°C |
Density | 0.8660g/mL |
Boiling Point | 111.0°C |
Quantity | 500 mL |
Flash Point | 4°C |
Packaging | EcoSafPak™ Glass Bottle |
Physical Form | Liquid |
Refractive Index | 1.4968 |
Assay Percent Range | 99.5% |
Residue after Evaporation | 0.001% max. |
Show More |
Safety and Handling
Product Identifier
- Toluene
- Danger
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P331-Do NOT induce vomiting.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'