missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toluene, ACS Reagent, ≥99.5%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Melting Point | -93°C (literature) |
CAS | 108-88-3 |
CAS Max % | 100.0000% |
Molecular Formula | C7H8 |
Linear Formula | C6H5CH3 |
MDL Number | MFCD00008512 |
UN Number | UN1294 |
Beilstein | 635760 |
Synonym | methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene |
InChI Key | YXFVVABEGXRONW-UHFFFAOYSA-N |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'