missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toluene, CHROMASOLV™, for HPLC, 99.9%, Honeywell Riedel-de Haen
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
2 L
2.5 L
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
missing translation for 'unitSize'
1 L
2 L
2.5 L
Pack of 4
Pack of 6
Beschreibung
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Spezifikation
Spezifikation
| Melting Point | -93°C (literature) |
| Absorbance | max. 1.00 at 286 nm, max. 0.01 at 400 nm, max. 0.01 at 350 nm, max. 0.10 at 300 nm, max. 0.50 at 288 nm |
| CAS Max % | 100.0000% |
| Density | 0.865 g/mL at 25 °C(lit.) |
| Boiling Point | 110.6°C |
| Quantity | 4 x 2.5 L |
| Flash Point | 4°C |
| Packaging | Glass Bottle |
| Linear Formula | C6H5CH3 |
| UN Number | UN1294 |
| Mehr anzeigen |
Sicherheit und Handhabung
Produktidentifikator
- Toluene
- Danger
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- Reproductive toxicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur