missing translation for 'onlineSavingsMsg'
Learn More
Toluene-d8, for NMR, 100.0 atom % D, packaged in 0.50 ml ampoules Product Code.: 10185030

Toluene-d8, for NMR, 100.0 atom % D, packaged in 0.50 ml ampoules

Product Code. 10185030
5 mL, Ampule
Click to view available options
Quantity:
5 mL
Packaging:
Ampule
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10185030

Brand: Thermo Scientific Acros 217440050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 2037-26-5 | C7H8 | 100.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Specifications

Density 0.9400g/mL
Flash Point 4°C
Packaging Ampule
Refractive Index 1.494
Quantity 5 mL
CAS 2037-26-5
Melting Point -84.0°C
Boiling Point 110.0°C
Molecular Formula C7H8
Linear Formula C6D5CD3
MDL Number MFCD00044638
Beilstein 05, IV, 797
Synonym toluene-d8, 2h8 toluene, perdeuteriotoluene, benzene-d5, methyl-d3, perdeuterotoluene, benzene-d5-, methyl-d3, 1,2,3,4,5-pentadeuterio-6-trideuteriomethyl benzene, toluene d8, toluene-d8, 99 atom % d, toluene-d8, 99.6 atom % d
InChI Key YXFVVABEGXRONW-JGUCLWPXSA-N
SMILES CC1=CC=CC=C1
IUPAC Name 1,2,3,4,5-pentadeuterio-6-(trideuteriomethyl)benzene
Molecular Weight (g/mol) 100.19
PubChem CID 74861
Formula Weight 100.21
Grade NMR
Chemical Name or Material Toluene-d8, 100.0 atom % D
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Toluene-d8
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Toluene-d8, for NMR, 100.0 atom % D, packaged in 0.50 ml ampoules >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.