Learn More
Toluene, Fluka™
Analytical standard, reference material
Brand: Honeywell Fluka R1592-5ML
Additional Details : CAS Number : 108-88-3 Weight : 0.00430kg
Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
- Expert Technical Support
Specifications
Toluene | |
108-88-3 | |
Colorless | |
92.14 | |
-95°C |
Glass Bottle | |
5 mL | |
C6H5CH3 | |
Analytical Standard | |
110.6°C |