Learn More
Toluene, HPLC Grade, 99.7% min, Thermo Scientific Chemicals
£48.30 - £158.00
Chemical Identifiers
CAS | 108-88-3 |
---|---|
Molecular Formula | C7H8 |
Molecular Weight (g/mol) | 92.14 |
MDL Number | MFCD00008512 |
InChI Key | YXFVVABEGXRONW-UHFFFAOYSA-N |
Synonym | methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene |
PubChem CID | 1140 |
ChEBI | CHEBI:17578 |
IUPAC Name | toluene |
SMILES | CC1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11338137
|
Thermo Scientific Alfa Aesar
022903.K2 |
1 L |
£48.30
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11318137
|
Thermo Scientific Alfa Aesar
022903.M1 |
4 x 1 L |
£158.00
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Toluene is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
108-88-3 | |
92.14 | |
YXFVVABEGXRONW-UHFFFAOYSA-N | |
1140 | |
toluene |
C7H8 | |
MFCD00008512 | |
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene | |
CHEBI:17578 | |
CC1=CC=CC=C1 |
Specifications
93°C | |
0.867 | |
4°C (40°F) | |
1.4967 | |
C7H8 | |
635760 | |
Miscible with ethanol,benzene,diethyl ether,acetone,chloroform,glacial acetic acid and carbon disulfide. Immiscible with water. | |
CC1=CC=CC=C1 | |
92.14 | |
CHEBI:17578 | |
HPLC |
108-88-3 | |
111°C | |
Liquid | |
99.7% min. | |
MFCD00008512 | |
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene | |
YXFVVABEGXRONW-UHFFFAOYSA-N | |
toluene | |
1140 | |
92.14 | |
Toluene |
Safety and Handling
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H336-H361d-H373
missing translation for 'dotInformation' : Hazard Class: 3; Packaging Group: II
missing translation for 'einecsNumber' : 203-625-9
missing translation for 'rtecsNumber' : XS5250000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only