missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toluene, Laboratory Reagent, ≥99.3%, Honeywell™
Click to view available options
Quantity:
1 L
2.5 L
10 L
20 L
6 x 1 L
Packaging:
Glass Bottle
Stainless Steel Can
Description
- ≥95% for general laboratory use.

Specifications
Specifications
Melting Point | -95°C |
Color | Colorless |
Density | 0.865g/mL at 25°C |
Boiling Point | 110.6°C |
Quantity | 2.5 L |
Explosion Limit | 0.07 |
Flash Point | 4°C (39.2°F) |
Ignition Point | 997°F |
Packaging | Glass Bottle |
Physical Form | Liquid |
Show More |
Safety and Handling



Signal Word
- Danger
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : toluene
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Toluene, Laboratory Reagent, ≥99.3%, Honeywell™ >
Spot an opportunity for improvement?Share a Content Correction