missing translation for 'onlineSavingsMsg'
Learn More

Tolylene 2,6-diisocyanate, 97%, Thermo Scientific Chemicals

Catalog Number 10164690
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
91-08-7
C9H6N2O2
174.16
MFCD00002010
RUELTTOHQODFPA-UHFFFAOYSA-N
2,6-diisocyanatotoluene, toluene-2,6-diisocyanate, 2,6-toluene diisocyanate, 2-methyl-m-phenylene diisocyanate, 2,6-tdi, m-tolylene diisocyanate, meta-tolylene diisocyanate, toluene 2,6-diisocyanate, benzene, 1,3-diisocyanato-2-methyl, 2,6-diisocyanato-1-methylbenzene
7040
CHEBI:53557
1,3-diisocyanato-2-methylbenzene
CC1=C(C=CC=C1N=C=O)N=C=O
This item is not returnable. View return policy
91-08-7
C9H6N2O2
174.16
MFCD00002010
RUELTTOHQODFPA-UHFFFAOYSA-N
2,6-diisocyanatotoluene, toluene-2,6-diisocyanate, 2,6-toluene diisocyanate, 2-methyl-m-phenylene diisocyanate, 2,6-tdi, m-tolylene diisocyanate, meta-tolylene diisocyanate, toluene 2,6-diisocyanate, benzene, 1,3-diisocyanato-2-methyl, 2,6-diisocyanato-1-methylbenzene
7040
CHEBI:53557
1,3-diisocyanato-2-methylbenzene
CC1=C(C=CC=C1N=C=O)N=C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 13.0°C
Color Colorless to Yellow
Density 1.2200g/mL
Boiling Point 246.0°C to 247.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H3(NCO)2
Refractive Index 1.5700 to 1.5720
Quantity 1 g
Beilstein 13, III, 291
Specific Gravity 1.22
Viscosity 3 mPa.s (25°C)
Formula Weight 174.16
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Tolylene 2, 6-diisocyanate, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tolylene 2,6-diisocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'