missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOMM40 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 5E2C5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66658-1-IG-20UL
This item is not returnable.
View return policy
Description
TOMM40 is the channel-forming subunit of the translocase of the mitochondrial outer membrane complex that is essential for protein import into mitochondria.Specifications
TOMM40 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O96008, Q75Q40, Q9QYA2 | |
Tomm40 | |
TOMM40 Fusion Protein Ag13065 | |
20 μL | |
Primary | |
Human, Porcine, Mouse, Rat | |
Antibody | |
IgG1 |
Western Blot | |
5E2C5 | |
Unconjugated | |
TOMM40 | |
C19orf1, D19S1177E, p38.5, PER EC1, PEREC1, Protein Haymaker, TOM40, TOMM40 | |
Mouse | |
Protein G | |
RUO | |
102165927, 10452, 308416, 53333 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction