missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOMM40L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA562375
This item is not returnable.
View return policy
Description
Immunogen sequence: VVNKVLSSHF QVAHTIHMSA LGLPGYHLHA AYAGDWQLSP TEVFPTVVGD M Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
Potential channel-forming protein implicated in import of protein precursors into mitochondria.Specifications
TOMM40L | |
Polyclonal | |
Unconjugated | |
TOMM40L | |
hypothetical protein FLJ12770; mitochondrial import receptor subunit TOM40B; Protein TOMM40-like; RGD1562006; RP11-297K8.10; Tom40B; TOMM40B; Tomm40l; tomm40l {ECO:0000250; translocase of outer mitochondrial membrane 40 homolog (yeast)-like; translocase of outer mitochondrial membrane 40 homolog B; translocase of outer mitochondrial membrane 40 homolog, like; translocase of outer mitochondrial membrane 40 homolog-like; translocase of outer mitochondrial membrane 40 homolog-like (yeast); translocase of outer mitochondrial membrane 40 like; translocase of outer mitochondrial membrane 40B; UniProtKB:Q969M1}; zgc:85715 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84134 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q969M1 | |
TOMM40L | |
Recombinant protein corresponding to Human TOMM40L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only