Learn More
Faster™ Top Box Kit for Decontamination of SafeFAST Elite Cabinets with 1.5 in. Camlock Outlet
Top box attaches to the top of the Safety Cabinet to allow for fumigation and comes with a 1.5 in. Camlock.
Brand: Faster™ FX0021247600
This item is not returnable.
View return policy
Description
- Comes with a Camlock to attach fumigation machine
- Specific to SafeFAST Elite Cabinets
- Prevents fumigant escaping into the laboratory
Specifications
SafeFAST Premium 212 | |
200 mm |