Learn More
Faster™ Top Box Kit for Decontamination of SafeFAST Elite Cabinets with 1.5 in. Camlock Outlet
Top box attaches to the top of the Safety Cabinet to allow for fumigation and comes with a 1.5 in. Camlock.
Brand:  Faster™ FX0021847600
 View more versions of this product
Product Code. 16288700
  • £1130.00 / Each
Explore more special offers
Description
Description
  • Comes with a Camlock to attach fumigation machine
  • Specific to SafeFAST Elite Cabinets
  • Prevents fumigant escaping into the laboratory
Specifications
Specifications
SafeFAST Elite, Classic, Premium 218
200 mm