missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Tora™ CCS Foam Insert
Click to view available options
Color: Black
Description
- Insert fills the gap between the eyewear and the wearer’s face, further reducing the likelihood of injury from projectiles and other debris
- Fits 3M™ Tora™ CCS Safety Glasses
- Foam insert offers extra comfort and protection from sweat, ideal for hot and humid conditions
Specifications
Certifications/Compliance | EN 166: 2001 |
Color | Black |
Product Type | Foam Insert |
For Use With (Equipment) | Tora™ CCS safety glasses |
No. per Pack | 5/Bag, 100/Pk. |
Material | Foam |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'