Learn More
Emma Safety Footwear Torino Safety Shoes
The formally styled business shoe that protects the feet from above and below with water-resistant leather uppers, zero penetration in-sole and a steel safety toe cap. Specifically designed for standing, walking and the occapional run.
Brand:  Emma Safety Footwear MM13509046
 View more versions of this product
Product Code. 13264454
  • £166.00 / 1 pair
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • Nitrile Rubber Heat Resistant Outsole
  • ESD Certified
  • Chrome-free lining
  • Sanitized
Specifications
Specifications
EN ISO 20345:2011, EN 12568:2010
Black
Hydro-Tec™ Sanitized Silver & Wellness leather
46
Laces
Torino
Leather
Steel