missing translation for 'onlineSavingsMsg'
Learn More

WTW™ Total Nitrogen Test Kits

Product Code. 10201595
Click to view available options
Range:
0.5 to 15 mg/L
0.5 to 15 mg/L N Total
10 to 150 mg/L
Unit Size:
0.15kg
0.25kg
Each
This item is not returnable. View return policy

Product Code. 10201595

Brand: WTW™ 252018

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Obtain precision results using the CombiCheck Total Nitrogen NTotal cell ready-made test kit for the determination of total nitrogen in waste water, drinking water, and surface water.

  • Variety of instrumentation for testing nitrogen in water
  • For waste water, drinking water, surface water
  • Nitrospectral Method
  • Photometric Test Methods
  • CombiCheck Total Nitrogen NTotal cell test kit
  • Safe determinations of COD and NTotal in water
TRUSTED_SUSTAINABILITY

Specifications

Analytes DMP
For Use With (Application) For waste water, drinking water, surface water
For Use With (Equipment) PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, spectroFlex photometers
Includes Certificates
No. of Tests 9
Range 0.5 to 15 mg/L
Sufficient For 25 Tests
Test Method Photometric
Type CombiCheck Total Nitrogen NTotal cell test kit
compliance-icons
Product Identifier
  • Nges 00613 .
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid, phosphoric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.